Please login to your online account to display your discounted pricing

Fisherbrand™ SureOne Ultra Micropoint Pipette Tip, Universal Fit


Offers a full line of universal fit ultra micropoint pipet tips

Manufacturer: fisherbrand™  H10296RSFIS

 View more versions of this product

Catalog No. 02-707-137

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Sterility Sterile
Product Type SureOne™ Ultra Tip
Packaging Hinged Rack, 10 x 96
Length (English) 1.7 in.
Length (Metric) 4.3cm
For Use With (Equipment) Ultra micropoint pipet
Color Natural
Volume (Metric) 0.1 to 10μL
Certifications/Compliance RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, DNA-free
Tip Style Micro Point