Please login to your online account to display your discounted pricing

Fisherbrand™ SureOne Thin Wall Micropoint Pipette Tips, Universal Fit


Offers a full line of universal fit thin wall micropoint pipet tips

Manufacturer: fisherbrand™  TTW110FIS

 View more versions of this product

Catalog No. 02-707-452

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type SureOne™ Tip
Volume (Metric) 1 to 200μL
Sterility Nonsterile
Color Yellow
Autoclavable Yes
Tip Style Nonfilter, SureOne
Material Polypropylene
Packaging Bag
Length (English) 2 in.
Length (Metric) 5cm
For Use With (Equipment) Standard pipette