Fisherbrand™ SureOne™ Reload Empty Racks
For use with Fisherbrand SureOne universal pipet tips
Supplier:  Fisherbrand™ T1000ERFIS
 View more versions of this product
Catalog No. 02-707-443
Description
Description
  • Inserts (not included) sit flush with rack, allowing easy multichannel pipetting
  • Notched clasp designed to prop lid open for autoclave venting
  • Sure stack lid rim prevents racks from sliding around when stacked and carted around a lab
  • Safe lock latch allows for one-handed opening and closing ability—won't pop open when dropped
  • Sure snap design securely holds insert in place, preventing it from being lifted out while pipetting
  • Flip-lid can be either a hinged or lift-off lid; hinged format does not prevent use of a multichannel pipet
Order Info
Inserts not included; must be ordered separately.
Specifications
Specifications
Empty Rack
Standard Pipette Tips (1250μL)