Please login to your online account to display your discounted pricing

Fisherbrand™ SureOne™ Micropoint Pipette Tips, Universal Fit


New, improved Fisherbrand SureOne Reload system offers a full line of micropoint universal fit pipet tips

Manufacturer: fisherbrand™  TA104RSFIS

 View more versions of this product

Catalog No. 02-707-437

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Product Type SureOne™ Micropoint Tip
Volume (Metric) 0.1 to 10μL
Sterility Sterile
Color Clear
Graduations 2μL
Tip Style Hinged Rack, Nonfilter, SureOne
Material Polypropylene
Packaging Hinged Rack
Length (English) 1.25 in.
Length (Metric) 3.17cm
Certifications/Compliance RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, DNA-free
For Use With (Equipment) Research grade pipetters