Please login to your online account to display your discounted pricing
Fisherbrand™ SureOne™ Filter Tip Reload Pipette Tips
Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA
Manufacturer: fisherbrand™  TFR140300FIS
 View more versions of this product
Catalog No. 02-707-479
  • / Pack of 960 Each
  • / Case of 5 Pack
Description & Specifications
Specifications
Product Type Reload Filter Tip
Material Polypropylene
Sterility Sterile
Color Clear
Tip Style Extended, Filter, Low Retention, Universal Fit
Packaging Rack
Autoclavable Yes
Volume (Metric) 30 to 300μL