Please login to your online account to display your discounted pricing

Fisherbrand™ SureOne™ Filter Tip Reload Pipette Tips


Sterile and free of RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin and DNA

Manufacturer: fisherbrand™  TFR11320FIS

 View more versions of this product

Catalog No. 02-707-476

  • / Pack of 960 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Material Polypropylene
Product Type Reload Filter Tip
Sterility Sterile
Color Clear
Autoclavable Yes
Tip Style Beveled, Filter, Low Retention, Universal Fit
Volume (Metric) 2 to 20μL
Packaging Rack
Accessories

Accessories for Catalog Number: 02707476

Item Mfr. No. Item Description Price Per Unit Quantity  
02-707-444
Fisherbrand™ SureOne Reload Empty Racks
fisherbrand™
T200ERFIS
For use with Fisherbrand SureOne universal pipet tips Pack of 10 Each for $83.31
Case of 5 Pack for $416.55