Please login to your online account to display your discounted pricing
Fisherbrand™ SureOne Beveled Pipette Tips, Universal Fit: 1-200μL
Offers a full line of universal fit beveled pipet tips
Manufacturer: fisherbrand™  T113G-FIS
 View more versions of this product
Catalog No. 02-707-448
  • / Pack of 1000 Each
  • / Case of 10 Pack
Description & Specifications
Specifications
Product Type SureOne™ Tip
Volume (Metric) 1 to 200μL
Sterility Nonsterile
Color Yellow
Graduations 10, 50, 100μL
Tip Style Beveled Ends
Packaging Bulk
Length (English) 2 in.
Length (Metric) 5cm
Certifications/Compliance RNase, DNase, DNA-free