Please login to your online account to display your discounted pricing
Fisherbrand™ SureOne Beveled Pipette Tips, Universal Fit: 1-200μL
Offers a full line of universal fit beveled pipet tips
Manufacturer: fisherbrand™  T113RNSFIS
 View more versions of this product
Catalog No. 02-707-416
  • / Pack of 960 Each
  • / Case of 5 Pack
Description & Specifications
Specifications
Sterility Sterile
Product Type SureOne™ Tip
Material Polypropylene
Packaging Hinged Rack, 10 x 96
Length (English) 2 in.
Length (Metric) 5cm
Graduations 10, 50, 100μL
Autoclavable Yes
Tip Style Beveled Ends
Certifications/Compliance RNase, DNase, ATP, pyrogen, bioburden, PCR inhibitors, endotoxin, DNA-free
Volume (Metric) 1 to 200μL
Color Clear