missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Super Friendly Air'It™ Aerosol Duster

Blows lint, dust and loose dirt away instantly
Manufacturer: Fisherbrand™ 9000800
Description
Blows lint, dust and loose dirt away instantly
Ideal for delicate, difficult-to-clean and inaccessible equipment.
- Provides high-pressure, pinpointed jets of air exactly where needed—up to 3000 jets of cleaning power
- Trigger-actuated valve provides controlled delivery at 71psi
- Triple-filtered (to 0.3μm) gas is noncorrosive, nonflammable, dry, and odorless
- 5 in. (13cm) snorkel constructed of nonconductive polyethylene
Certifications
EPA designated as ozone-safe
Specifications
11 oz. | |
11 oz., 312g | |
Ideal for delicate, difficult-to-clean and inaccessible equipment | |
Super Friendly Air'It™ aerosol duster |
312g | |
No | |
71psi |