Please login to your online account to display your discounted pricing

Fisherbrand™ Straight Dropper


Manufacturer: fisherbrand™  LF-011

 View more versions of this product

Catalog No. S12345

  • / Pack of 144 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Capacity (Metric) 2mL
Capacity (English) 0.06 oz.
Material Glass
Shape Straight
Capacity 0.06 oz., 2mL