Please login to your online account to display your discounted pricing
Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
Choose skirted or conical-bottom style
Manufacturer: fisherbrand™  512SFIS
 View more versions of this product
Catalog No. 02-681-371
  • / Pack of 500 Each
  • / Case of 5 Pack
Description & Specifications
Specifications
Max. Force 25000xG
Sterile Yes
Color Natural
Capacity (Metric) 0.5mL
Shape Conical
Closure Material Polypropylene
Closure Type Screw
Includes O-ring
Material Polypropylene
Accessories
Accessories for Catalog Number: 02681371
Item Mfr. No. Item Description Price Per Unit Quantity  
05-407-22
Fisherbrand™ Microcentrifuge Tube Accessory Micro Tube Tool
fisherbrand™
BIP4000
For use in capping or decapping Fisherbrand Microcentrifuge Tubes with Snap Caps Pack of 4 Each for $14.20
Case of 25 Pack for $355.00