Fisherbrand™ Sterile Microcentrifuge Tubes with Screw Caps
Choose skirted or conical-bottom style
Manufacturer:  Fisherbrand™ 522SFIS
 View more versions of this product
Catalog No. 02-681-375
Specifications Description & Specifications
Specifications
Polypropylene
Yes
Natural
25000xG
O-ring
Screw
2mL
Polypropylene
Yes
Conical
Accessories