Please login to your online account to display your discounted pricing

Fisherbrand™ Standardization Pipet Tips


Eliminate the need to buy pipetter-specific tips.

Manufacturer: fisherbrand™  A22220FIS/EMD

 View more versions of this product

Catalog No. S98754B

  • / Pack of 750 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Standardization Tip
Volume (Metric) 200 to 1000μL
Sterility Sterile
Color Natural
Autoclavable Yes
Tip Style Universal Fit
Material Polypropylene
Packaging Bulk
Length (English) 3.4 in.
Length (Metric) 8.7cm
Certifications/Compliance RNase, DNase, DNA, PCR inhibitor, pyrogen-free
For Use With (Equipment) Research grade pipetters