Please login to your online account to display your discounted pricing

Fisherbrand™ Standardization Pipet Tips


Eliminate the need to buy pipetter-specific tips.

Manufacturer: fisherbrand™  A22070FIS/EMD

 View more versions of this product

Catalog No. S98754A

  • / Pack of 1000 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Standardization Tip
Volume (Metric) 0 to 200μL
Sterility Sterile
Color Natural
Autoclavable Yes
Tip Style Universal Fit
Material Polypropylene
Packaging Bulk
Length (English) 2 in.
Length (Metric) 5cm
Certifications/Compliance RNase, DNase, DNA, PCR inhibitor, pyrogen-free
For Use With (Equipment) Research grade pipetters