Please login to your online account to display your discounted pricing

Fisherbrand™ Standardization Pipet Tips: 500–1300μL


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  A23191FIS

 View more versions of this product

Catalog No. 02-681-177

  • / Pack of 1000 Each
  • / Case of 5 Pack
 

Description & Specifications

Specifications

Volume (Metric) 500 to 1300μL
Length (Metric) 7.9cm
For Use With (Equipment) Research grade pipetters
Length (English) 3.13 in.
Product Type Standardization Tip
Sterility Sterile
Material Polypropylene
Color Natural
Autoclavable Yes
Packaging Lift-off Rack, 10 x 100
Certifications/Compliance RNase, DNase, DNA, bioburden, PCR inhibitor and pyrogen free
Tip Style Universal Fit