Please login to your online account to display your discounted pricing

Fisherbrand™ Standardization Pipet Tips: 200–1000μL


Premium virgin polypropylene resin and advanced molding methods produce a superior, high-quality pipet tip

Manufacturer: fisherbrand™  A22221FIS

 View more versions of this product

Catalog No. 02-681-161

  • / Pack of 600 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type Standardization Tip
Volume (Metric) 0 to 1000μL
Sterility Sterile
Color Natural
Autoclavable Yes
Tip Style Universal Fit
Material Polypropylene
Packaging Stack Rack
Length (English) 3.4 in.
Length (Metric) 8.7cm
Certifications/Compliance RNase, DNase, DNA, PCR inhibitor, pyrogen-free
For Use With (Equipment) Research grade pipetters