Fisherbrand™ Standard-Top Serological Pipets
Provide fast, uniform flow times.
Manufacturer:  Fisherbrand™ 13675J
 View more versions of this product
Catalog No. 13-675J
Description
Description
Provide fast, uniform flow times.
Accuracy printed on each pipet
  • Borosilicate glass, permanent amber markings
  • Tuf-Temp™ treated to reduce chipping
  • All Fisherbrand Reusable Serological Pipets also available as PersonaPet™ personalized pipets
  • Pipets 13-675D and 13-675E are recommended for VDRL test
Specifications
Specifications
±0.02mL
0.06 oz., 2mL
White
Yes
Standard Pack
2mL
0.06 oz.
ASTM E1044-96
White
0.01mL
Serological Pipet
Borosilicate Glass