Please login to your online account to display your discounted pricing

Fisherbrand™ Standard Disposable Transfer Pipettes

Single squeeze draws up to 3.2mL

$45.81 - $56.19

Specifications

Bulb Draw (Metric) 3.2mL
Disposable Yes
For Use With (Application) Hematology, urinalysis, chemistry, Blood banking
Material Polyethylene
View More Specs
Products
Catalog Number Mfr. No. Graduated Capacity (Metric) Length (Metric) Price Quantity    

13-711-7M

 
fisherbrand™
13-711-7M
No 7.7mL 15cm Pack of 500 Each for $45.81
Case of 10 Pack for $458.10

13-711-9D

 
fisherbrand™
SAM225
Yes 7.5mL 15.5cm Pack of 500 Each for $56.19
Case of 10 Pack for $561.90
Description & Specifications

Specifications

Bulb Draw (Metric) 3.2mL
Disposable Yes
For Use With (Application) Hematology, urinalysis, chemistry, Blood banking
Material Polyethylene

Unbreakable all-in-one pipettes eliminate the hazards of broken glass and exposure to infectious materials.

  • Put an end to matching rubber bulb with glass pipette. Since entire unit is disposable, these pipettes prevent cross-contamination to samples and reduce airborne contamination to personnel
  • One-piece design molded from see-through, low-density polyethylene
  • Inert to biological fluids and most acids
  • Work well in hematology, urinalysis, chemistry, blood banking — anywhere there is a need for quick, safe transfer of fluids
  • In four ready-to-use styles
  • One squeeze draws from 2.0 to 3.3mL (depending on style used)
  • All deliver repeatable drop sizes
  • Can be sealed and refrigerated for sample storage (seal with Cat. No. 02-678 Hemato-Seal™ Tube Sealant)

  • 13-711-9D has 0.50mL graduations

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.