Please login to your online account to display your discounted pricing

Fisherbrand™ Stainless Steel Clamps


Excellent chemical resistance and overall strength

Manufacturer: fisher scientific company  916405

 View more versions of this product

Catalog No. 02-217-005

  • / Each
 

Description & Specifications

Specifications

Product Type Jumbo Holder
Size Jumbo
Size (English) Grip 0.83 in.
Size (Metric) Grip 2.1cm
For Use With (Application) Secures clamps to lab frames, accepts rods up to 0.83 in.
Material Stainless Steel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.