Fisherbrand™ Squibb Pear-Shaped Separatory Funnels with PTFE-Plug Stopcock
Do not require grease
Manufacturer:  Fisherbrand™ KFS29048F125
 View more versions of this product
Catalog No. 13-678-603
Description
Description
Do not require grease
Lower stems made of large-diameter tubing for liquid to drain out completely when stopcock is open.
  • KIMAX glass
  • Eliminate contamination
  • Will not bind or freeze
Order Info
Stem lengths, 60mm.
Compliance
Designed from ASTM™ E 1096-86.
Specifications
Specifications
ASTM E1096, Type IV
60mm
6 Case
Glass
125mL
Borosilicate Glass
Eliminates contamination
Squibb Separatory Funnel
2; PTFE
22
222mm