Fisherbrand™ Split Cast-Brass Rings
Hold leveling bulbs safely with these cast-brass rings
$41.66 - $41.66
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) Outer | Diameter (Metric) Outer | Price | Quantity | ||||
14-052AQ
|
Fisherbrand™
00440Q |
2 in. | 5.1cm |
Each for $41.66
|
|
Description
- Split ring allows easy placement and removal of bulb
- Vinyl-coated sheath on ring protects bulb from breakage Integral clamp holds ring tightly to support rods