Please login to your online account to display your discounted pricing
Fisherbrand™ Special Test Papers
Lead acetate paper and potassium iodide/starch paper for testing presence of soluble sulfides and chlorine levels, respectively
$63.20 - $67.71
Specifications
For Use With (Application) For testing presence of soluble sulfides and chlorine levels, respectively
Quantity 24 Pack
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
14-860
precision laboratories
16024V100
Potassium Iodide/Starch Test Strip Pack of 24 Each for $67.71
14-862
precision laboratories
PRL175
Lead Acetate Test Paper Pack of 24 Each for $63.20
Description & Specifications
Specifications
For Use With (Application) For testing presence of soluble sulfides and chlorine levels, respectively
Quantity 24 Pack
  • Lead Acetate paper turns black in presence of hydrogen sulfide and other soluble sulfides
  • Potassium Iodide/Starch paper turns blue to deep purple in the presence of low levels of chlorine, iodine, peroxide and ozone
100 Strips per vial