Please login to your online account to display your discounted pricing
Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
Are pre-printed with Chemical name and formula. Bel Art™ Solvent-Vented™ and Safety-Labeled Wash Bottles have patented valve eliminates drips and allows bottle to dispense upright or inverted.
$53.72 - $55.64
Specifications
Material LDPE
Closure Material Polypropylene
Autoclavable No
Color Translucent
Certifications/Compliance OSHA 29 CFR 1910.1200
View More Specs
Products
Catalog Number Mfr. No. For Use With (Application) Price Quantity    
03-409-31
  Accessories
fisherbrand™
116423611
Methyl Ethyl Ketone Pack of 3 Each for $53.72
03-409-35
  Accessories
fisherbrand™
116423624
Isopropanol Pack of 3 Each for $53.72
03-409-37
  Accessories
fisherbrand™
116423622
Acetone Pack of 3 Each for $53.72
03-409-30
  Accessories
fisherbrand™
116423612
Ethyl Acetate Pack of 3 Each for $53.72
03-409-33
  Accessories
fisherbrand™
116423623
Methanol Pack of 3 Each for $53.72
03-409-34
  Accessories
fisherbrand™
116423638
Unlabeled Pack of 3 Each for $53.72
03-409-36
  Accessories
fisherbrand™
116423639
Ethanol Pack of 3 Each for $55.64
Description & Specifications
Specifications
Material LDPE
Closure Material Polypropylene
Autoclavable No
Color Translucent
Certifications/Compliance OSHA 29 CFR 1910.1200
Mouth Wide
Capacity (Metric) 500mL
  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Prelabeled with six common solvents; also available unlabeled.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.