Please login to your online account to display your discounted pricing

Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles


Are pre-printed with Chemical name and formula. Bel Art™ Solvent-Vented™ and Safety-Labeled Wash Bottles have patented valve eliminates drips and allows bottle to dispense upright or inverted.

Manufacturer: fisherbrand™  116423639

 View more versions of this product

Catalog No. 03-409-36

  • / Pack of 3 Each
 

Description & Specifications

Specifications

For Use With (Application) Ethanol
Mouth Wide
Autoclavable No
Capacity (English) 16 oz.
Certifications/Compliance OSHA 29 CFR 1910.1200
Material LDPE
Color Translucent
Capacity (Metric) 500mL
Capacity 16 oz., 500mL
Closure Material Polypropylene

  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available

 

Use LYOB only with solvents compatible with LDPE.


Accessories

Accessories for Catalog Number: 0340936

Item Mfr. No. Item Description Price Per Unit Quantity  
03-409-24
Bel-Art™ SP Scienceware™ Wash Bottle Carrier
bel-art™
F116400001
Transports up to 6 wash bottles. Bel Art™ SP Scienceware™ Wash Bottle Carrier offers safe, convenient, lightweight transportation that is sturdy and lightweight. Each for $159.14