Fisherbrand™ Solvent-Vented™ and Safety-Labeled Wash Bottles
Are pre-printed with Chemical name and formula. Bel Art™ Solvent-Vented™ and Safety-Labeled Wash Bottles have patented valve eliminates drips and allows bottle to dispense upright or inverted.
Manufacturer:  Fisherbrand™ 116423624
 View more versions of this product
Catalog No. 03-409-35
Specifications
Description
Bottle, Wash; Fisherbrand; Solvent-vented; LDPE; Wide-mouth; Color-coded PP closures; Safety-Labeled DOT, NFPA, CAS protective euip and target organ; easy to read; 16 oz. (500mL); Isopropanol
  • Wide mouth 2 in. (53mm) bottle for easy refilling
  • Polypropylene closure with leakproof design, color-coded to aid in easy identification
  • Translucent, LDPE solvent-resistant; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • Labeled with six of the most commonly used solvent names or label your own; custom labeling available
Use LYOB only with solvents compatible with LDPE.
Description & Specifications
Specifications
16 oz., 500mL
16 oz.
Polypropylene
Isopropanol
No
500mL
OSHA 29 CFR 1910.1200
Translucent
LDPE
Wide
Accessories