Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ D1069810
 View more versions of this product
Catalog No. 09-704-1F
Specifications Description & Specifications
Specifications
2cm
0.55 in.
18D
0.787 in.
Translucent
1.4cm
Ideal for procedures demanding high purity ideal for biological applications
-76° to 428°F
1.8cm
Solid Stopper
Silicone
-60° to 220°C
0.708 in.
1/18D