Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ D1069807
 View more versions of this product
Catalog No. 09-704-1C
Specifications Description & Specifications
Specifications
2cm
0.5cm
Ideal for procedures demanding high purity ideal for biological applications
-76° to 428°F
1.2cm
0.787 in.
000/13D
Solid Stopper
-60° to 220°C
13D
Silicone
0.196 in.
0.472 in.
Translucent