Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ D1069816
 View more versions of this product
Catalog No. 09-704-1L
Specifications Description & Specifications
Specifications
1.14 in.
-76° to 428°F
1.37 in.
34D
2.9cm
Ideal for procedures demanding high purity ideal for biological applications
3cm
-60° to 220°C
Solid Stopper
1.18 in.
3.5cm
Translucent
Silicone
6.5/34D