Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ D1069813
 View more versions of this product
Catalog No. 09-704-1I
Specifications Description & Specifications
Specifications
0.826 in.
27D
2.1cm
1.06 in.
4/27D
2.7cm
Ideal for procedures demanding high purity ideal for biological applications
1.18 in.
Silicone
-76° to 428°F
Translucent
Solid Stopper
-60° to 220°C
3cm