Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ DX263029-10
 View more versions of this product
Catalog No. 09-704-1J
Specifications Description & Specifications
Specifications
2.3cm
Ideal for procedures demanding high purity ideal for biological applications
29D
Solid Stopper
0.905 in.
Silicone
5/29D
3cm
1.18 in.
-60° to 220°C
1.141 in.
-76° to 428°F
2.9cm
Translucent