Fisherbrand™ Solid Silicone Stoppers
Create a tight, reliable seal in virtually all sealing applications
Manufacturer:  Fisherbrand™ D1069823
 View more versions of this product
Catalog No. 09-704-1T
Specifications Description & Specifications
Specifications
-60° to 220°C
2.2 in.
Ideal for procedures demanding high purity ideal for biological applications
-76° to 428°F
Silicone
65D
11.5/165D
4.5cm
5.6cm
1.771 in.
6.5cm
Solid Stopper
2.55 in.
Translucent