Please login to your online account to display your discounted pricing

Fisherbrand™ Soil Analysis ASTM Hydrometers


These hydrometers are used to determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)

Manufacturer: fisher scientific  B618913120

 View more versions of this product

Catalog No. 13-202-132

  • / Each
 

Description & Specifications

Specifications

Recommended Storage Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Accuracy ±0.001
Specific Gravity Range 0.995 to 1.038
For Use With (Application) To determine the specific gravity of soil in the range of 0.995 to 1.038 or -5 to 60g/L (using a weight by volume scale)
Certifications/Compliance Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team in exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC /MRA and located in ISO 14001:2004 Environmental Management System registered facility.
Length (Metric) 280mm
ASTM Number 151H
Increments 0.001
Accessories

Soil analysis hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.