missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ Soft Glass Rods
With rounded and fire-polished ends.
Manufacturer: Fisherbrand™ S63450
Description
- Constructed of glass
- A stirring rod is used to mix chemicals and liquids for laboratory purposes
Specifications
250mm | |
Case of 1 pack (72 rods/pk.) | |
Rounded and fire polished ends | |
6mm |
Glass | |
0.23 in. | |
9.84 in. |