Fisherbrand™ Snap-Cap™ Microcentrifuge Tubes: Flat Top
Polypropylene tubes
Manufacturer:  Fisherbrand™ 502PLNOFIS
 View more versions of this product
Catalog No. 02-681-245
Description
Description
  • Packed in bulk Nonsterile
Specifications
Specifications
Snap Cap with Flat Top
RNase, DNase, RNA, DNA, ATP, pyrogen-free
0.6mL
10mm
Polypropylene
Yes
Non-sterile
Snap
Orange
30mm
30000xG
Bulk
Documents