Please login to your online account to display your discounted pricing

Fisherbrand™ Single Cytology Funnels


For the Thermo Scientific™ Shandon™ CytoSpin™ Centrifuge

Manufacturer: fisherbrand™  10-355*

 View more versions of this product

Catalog No. 10-355

  • / Case of 500 Each
 

Description & Specifications

Specifications

For Use With Shandon CytoSpin Centrifuge
Color White
Quantity 500/Cs.
Volume (Metric) Up to 0.4mL or less