Please login to your online account to display your discounted pricing

Fisherbrand™ Single Cytology Funnels


For the Thermo Scientific™ Shandon™ CytoSpin™ Centrifuge

Manufacturer: fisherbrand™  10-354

 View more versions of this product

Catalog No. 10-354

  • / Pack of 50 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Includes White Filter Card
Volume (Metric) Up to 0.4mL
Color White
For Use With Shandon CytoSpin Centrifuge
Quantity 50/Pk.