Bio Plas Siliconized Screw-Cap Microcentrifuge Tubes
Ideal for packaging, transporting, and storing diagnostic tests, especially in the area of biotechnology, molecular biological products and reagents
Manufacturer:  Bio Plas 4204SL
 View more versions of this product
Catalog No. 05-541-66
Description
Description
Low surface adhesion reduces samples adhering to walls.
  • Polypropylene
  • Natural color
  • Conical with skirt
  • Available sterile or nonsterile, with or without screw cap
  • Withstand centrifugal forces up to 25,000×G
  • Sterile model includes tight-seal closure that reduces accidental opening during use
Order Info
1000 per pack, 5 packs per case
Specifications
Specifications
Non-sterile
2mL
Polypropylene
Conical
Without
Natural
25000xG
Documents