Please login to your online account to display your discounted pricing

Silicone Recessed Septum Stoppers


Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal

Manufacturer: fisher scientific  FB57897

 View more versions of this product

Catalog No. FB57897

  • / Pack of 10 Each
 

Description & Specifications

Specifications

For Use With (Application) Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
Diameter (Metric) Inside 14.50 to 15.50mm
For Use With (Equipment) For 0.145 to 0.155cm I.D. containers
Product Type Silicone Recessed Septum Stopper
Size 1
Stopper Number 1
Material Silicone
Color Translucent