Fisherbrand™ Silicone Recessed Septum Stoppers
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
Description
- Elliptical flange to allow efficient removal of stopper from aperature
- Easily pierced with a hypodermic needle
Specifications
Translucent | |
0 | |
Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal | |
Silicone |
Silicone Recessed Septum Stopper | |
13.75 to 14.25 mm | |
0 | |
For 13.75 to 14.25mm I.D. containers |