Please login to your online account to display your discounted pricing

Silicone Recessed Septum Stoppers


Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal

Manufacturer: fisher scientific  FB57895

 View more versions of this product

Catalog No. FB57895

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Diameter (Metric) Inside 9.22 to 9.75mm
For Use With (Application) Nontoxic and autoclavable recessed septum stoppers are externally serrated to create an effective internal seal
For Use With (Equipment) For 0.922 to 0.975cm I.D. containers
Color Translucent
Material Silicone
Product Type Silicone Recessed Septum Stopper