Please login to your online account to display your discounted pricing

Fisherbrand™ Silicone Plate Seals


Excellent chemical compatibility and resistance to tearing and coring

Manufacturer: fisherbrand™  0339656

 View more versions of this product

Catalog No. 03-396-56

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Width (Metric) 8.6mm
Pre-slit Yes
Product Type 384 Well Silicone Plate Seal
Autoclavable Yes
Color Clear
No. per Pack 10.0
Applications Excellent chemical compatibility and resistance to tearing and coring
Material Silicone