Please login to your online account to display your discounted pricing

Fisherbrand™ Silicone Plate Seals


Excellent chemical compatibility and resistance to tearing and coring

Manufacturer: fisherbrand™  0339655

 View more versions of this product

Catalog No. 03-396-55

  • / Pack of 10 Each
 

Description & Specifications

Specifications

No. per Pack 10.0
Width (Metric) 8.6mm
Material Silicone
Applications Excellent chemical compatibility and resistance to tearing and coring
Pre-slit No
Autoclavable Yes
Product Type 384 Well Silicone Plate Seal
Color Clear