Fisherbrand™ Silicone Plate Seals
Provides excellent chemical compatibility and resistance to tearing and coring.
Manufacturer:  Fisherbrand™ 0339653
 View more versions of this product
Catalog No. 03-396-53
Description
Description
  • Nonslit seals reseal after multiple penetrations to prevent sample evaporation and cross-well contamination
  • Preslit seals are easy to penetrate and reduce well vacuum
  • Apply by hand or with an applicator
  • Remain flexible at ambient temperatures and in the absence of chlorinated solvents
  • Autoclavable
Specifications
Specifications
Silicone
Clear
96 Well Square Silicone Plate Master Block Seal
No
Excellent chemical compatibility and resistance to tearing and coring
10.0
Yes
8.6mm
Documents