Please login to your online account to display your discounted pricing

Fisherbrand™ Silicone Plate Seals


Excellent chemical compatibility and resistance to tearing and coring

Manufacturer: fisherbrand™  0339650

 View more versions of this product

Catalog No. 03-396-50

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Material Silicone
No. per Pack 10.0
Product Type 96 Well Silicone Plate Seal
Autoclavable Yes
Pre-slit Yes
Width (Metric) 7mm
Applications Excellent chemical compatibility and resistance to tearing and coring
Color Clear