Fisherbrand™ Silicone Bath Fluids
Provide thermal stability over a variety of temperature ranges.
$2,551.87 - $2,551.87
Products 1
Catalog Number Mfr. No. Type Viscosity Temperature Range Price Quantity  
Catalog Number Mfr. No. Type Viscosity Temperature Range Price Quantity  
13-874-60B
View Documents Promotion Details
Fisherbrand™
2101
Heat Transfer Fluid 510 14 cs. 0° to 88°C
Each for $2,551.87
Description
Description
  • Suitable for use in stainless-steel reservoirs
  • Supplied in 1 gal. (3.8L) container
SDS