Fisherbrand™ Silicone Bath Fluids
Provide thermal stability over a variety of temperature ranges
Manufacturer:  Fisherbrand™ 2201*
 View more versions of this product
Catalog No. 13-874-60C
Description
Description
  • Suitable for use in stainless-steel reservoirs
  • Supplied in 1 gal. (3.8L) container
Specifications
Specifications
DC550
Silicone
Silicone Bath Fluid
100°C to 200°C
-50°C
1 gal.
125 Cs.
Documents