Fisherbrand™ Sharps-A-Gator™ Sharps Containers
Puncture-resistant with wide rectangular opening for horizontal loading of needles
Manufacturer:  Fisherbrand™ 31156916
 View more versions of this product
Catalog No. 22-037-959
Description
Description
Puncture-resistant with wide rectangular opening for horizontal loading of needles
  • General-purpose sharps container with clear locking lid and marked with biohazard label
  • Made from puncture-resistant polypropylene that can be autoclaved or incinerated
  • Easily mount on walls and carts, nest to conserve storage space
Compliance
Used to comply with OSHA standards for sharps and biohazard waste collection.
Specifications
Specifications
Autoclavable
Used to comply with OSHA standards for sharps and biohazard waste collection.
Slide
7 x 10.5 x 10.25 in. (18 x 27 x 26 cm)
10.25 in.
7 in.
20/Cs.
27 cm
Red
7.5 L
Clear
Rectangle
Yes
26 cm
18 cm
10.5 in.
2 gal.
Puncture-Resistant Polypropylene
Documents