missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Sharps-A-Gator™ Sharps Containers

Puncture-resistant with wide rectangular opening for horizontal loading of needles

Manufacturer:  Fisherbrand™ 31157160

 View more versions of this product

Catalog No. 14-827-71




Description

Description

Puncture-resistant with wide rectangular opening for horizontal loading of needles

  • General-purpose sharps container with clear locking lid and marked with biohazard label
  • Made from puncture-resistant polypropylene that can be autoclaved or incinerated
  • Easily mount on walls and carts, nest to conserve storage space

Compliance

Used to comply with OSHA standards for sharps and biohazard waste collection.

Specifications

Specifications

38 L
Clear
12 x 15.5 x 21.5 in. (31 x 40 x 55 cm)
21.5 in.
12 in.
6/Cs.
15.5 in.
Autoclavable
Red
Used to comply with OSHA standards for sharps and biohazard waste collection.
Lockable
Yes
55 cm
31 cm
Rectangle
40 cm
10 gal.
Polypropylene
SDS
Documents