missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Serialized Class A Cylinders with Certificate of Traceability

Calibrated “To Contain”

Manufacturer:  Fisherbrand™ 08559D

 View more versions of this product

Catalog No. 08-559D



 

Specifications Description & Specifications

Specifications

260mm
A
Traceable NIST certificate
±0.5mL
1mL
Borosilicate Glass
100mL
White scale
10.2 in.
1mL
Accessories