missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Serialized Class A Cylinders with Certificate of Traceability

Calibrated “To Contain”

Manufacturer:  Fisherbrand™ 08559C

 View more versions of this product

Catalog No. 08-559C



 

Specifications Description & Specifications

Specifications

190mm
A
Traceable NIST certificate
±0.25mL
1mL
Borosilicate Glass
50mL
White scale
7.48 in.
1mL
Accessories